Sorbic acid ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:110-44-1
:
Formula:CH₃CH=CHCH=CHCOOH
MDL Number:MFCD00002703
:112.13 g/mol
Merck Index:14,08721
Boiling Pt:228°C
Melting Pt:132…135 °C
Density:1,204 g/cm³ (20 °C)
Flash Pt: (closed cup)
EINECS:203-768-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:133.0 to 136.0 deg-C
- Solubility in Acetone:very faint turbidity
Specification Test Results
Frequently Bought Together