
Tetraethylammonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:71-91-0
:
Formula:C₈H₂₀BrN
MDL Number:MFCD00011825
:210.16 g/mol
Merck Index:14,09199
Melting Pt:285°C
Density:1,43 g/cm³ (20 °C)
EINECS:200-769-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together