Order Entry
Netherlands
ContactUsLinkComponent
Polyethylene glycol 400
Polyethylene glycol 400
Supplier:  TCI
Certificates
Polyethylene glycol 400
Supplier:  TCI
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Colorless to Almost colorless clear liquid
  • Average molecular weight:
    380 to 420