

Pentafluorobenzoyl chloride ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2251-50-5
:
Formula:C₇ClF₅O
MDL Number:MFCD00000657
:230.52 g/mol
Boiling Pt:159°C
Density:1,6 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:218-842-4
UN:3265
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Effect test of GC derivalization:Effective as derivatization agent for 1-Pentadecanol
Specification Test Results
Related Information
Frequently Bought Together