

Isopropyl formate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:625-55-8
:
Formula:C₄H₈O₂
MDL Number:MFCD00014127
:88.11 g/mol
Boiling Pt:68°C
Melting Pt:–89 °C
Density:0,8826 g/cm³ (0 °C)
Flash Pt:–6°C
EINECS:210-901-2
UN:1281
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together