Order Entry
Netherlands
ContactUsLinkComponent
Sodium formate ≥97.0% (by titrimetric analysis)
Sodium formate ≥97.0% (by titrimetric analysis)
  TCIAS0807-500G
 :  TCI
 :  
undefined
Sodium formate ≥97.0% (by titrimetric analysis)
  TCIAS0807-500G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 97.0 %

 

 

 :
 :68.01 g/mol
 :14,08621
 :253°C
 :1,919 g/cm³ (20 °C)
 :Ambient
 :205-488-0
 :01-2119486468-21
 :141-53-7
 :HCO₂Na
 :MFCD00013101