Order Entry
Netherlands
ContactUsLinkComponent
4-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
Irritant
4-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAF0560-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAF0560-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    165.0 to 169.0 deg-C

Specification Test Results

About this item

 :
 :154.14 g/mol
 :GHS07
CAS Number:403-15-6
Formula:C₈H₇FO₂
MDL Number:MFCD01631424