
3-Bromo-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:176548-70-2
:
Formula:C₇H₄BrFO₂
MDL Number:MFCD01569540
:219.01 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:142.0 to 146.0 deg-C
Specification Test Results
Frequently Bought Together