Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2-fluoropyridine
3-Bromo-2-fluoropyridine ≥98.0% (by GC)
3-Bromo-2-fluoropyridine ≥98.0% (by GC)
:
TCI
3-Bromo-2-fluoropyridine ≥98.0% (by GC)
:
TCI
:
36178-05-9
:
:
C₅H₃BrFN
:
MFCD04112496
:
175.99 g/mol
:
76°C
:
6.1,III
:
GHS07
Appearance:
Colorless to Light yellow clear liquid
Purity (GC):
min. 98.0 %
Manual Fluorination Reagents, Fluorinated Building Blocks
Frequently Bought Together
Tetrachloroethylene, EMPLURA®, Supelco®
: Merck
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Butyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Glycerine 85% Ph. Eur.
: VWR Chemicals
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Toluene, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Methanol, SupraSolv® for gas chromatography, Supelco®
: Merck
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals