
Methyl-3-aminobenzoate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4518-10-9
:
Formula:C₈H₉NO₂
MDL Number:MFCD00017102
:151.16 g/mol
Boiling Pt:170°C
Melting Pt:148…152 °C
Density:1,232 g/cm³ (20 °C)
EINECS:224-842-5
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:53.0 to 57.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together