
α,α,α-Trifluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:454-92-2
:
Formula:C₈H₅F₃O₂
MDL Number:MFCD00002519
Boiling Pt:126°C
Melting Pt:106 °C
Density:1,371 g/cm³ (25 °C)
:190.12 g/mol
Storage Temperature:Ambient
EINECS:207-230-2
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
- Melting point:104.0 to 107.0 deg-C
Specification Test Results
Frequently Bought Together