Order Entry
Netherlands
ContactUsLinkComponent
3,3-Diphenylpropionic acid ≥97.0% (by titrimetric analysis)
Irritant
3,3-Diphenylpropionic acid ≥97.0% (by titrimetric analysis)
Catalog # TCIAD1779-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3,3-Diphenylpropionic acid ≥97.0% (by titrimetric analysis)
Catalog # TCIAD1779-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    152.0 to 156.0 deg-C

Specification Test Results

About this item

 :
 :226.27 g/mol
Boiling Pt:365 °C (1013 hPa)
Melting Pt:156 °C
EINECS:210-125-4
 :GHS07
CAS Number:606-83-7
Formula:C₁₅H₁₄O₂
MDL Number:MFCD00002717