Bromocresol green
Supplier: TCI
Certificates
About this item
CAS Number:76-60-8
:
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874
:698.02 g/mol
Merck Index:14,01386
Melting Pt:217…218 °C
Storage Temperature:Ambient
EINECS:200-972-8
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Absorbance:min. 0.250 (5mg/L, pH5.8, 613.0 to 617.0 nm)
- Solubility in Sodium hydroxide solution:almost transparency
- Drying loss:max. 1.0 %
- Ignition residue (Sulfate):max. 0.5 %
Specification Test Results
Frequently Bought Together