2-Tetradecanone ≥97.0% (by GC)
TCIAT1314-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Melting point:33.0 to 37.0 deg-C
:
:212.38 g/mol
:148°C
:Ambient
:219-063-2
:2345-27-9
:C₁₄H₂₈O
:MFCD00060927