Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-Chloro-5-hydroxypyridine ≥97.0% (by GC, titration analysis)
  TCIAC2271-5G
 :  TCI
 :  
undefined
 
2-Chloro-5-hydroxypyridine ≥97.0% (by GC, titration analysis)
  TCIAC2271-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %

 

 

 :
 :129.55 g/mol
 :159°C
 :GHS07
 :41288-96-4
 :C₅H₄ClNO
 :MFCD00234050