
Methyl anthranilate ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:134-20-3
:
Formula:C₈H₉NO₂
MDL Number:MFCD00007710
:151.16 g/mol
Merck Index:14,06020
Boiling Pt:237 °C (1013 hPa)
Melting Pt:24 °C
Density:1,16 g/cm³ (20 °C)
EINECS:205-132-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
Specification Test Results
Frequently Bought Together