Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,7-Dihydroxynaphthalene ≥99.0% (by GC)
 :  TCI
 
undefined
 
2,7-Dihydroxynaphthalene ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (GC):
    min. 99.0 %

 

 

Frequently Bought Together