
Pyridine-2,6-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:499-83-2
:
Formula:C₇H₅NO₄
MDL Number:MFCD00006299
:167.12 g/mol
Boiling Pt:357 °C (1013 hPa)
Melting Pt:252°C
Flash Pt:188 °C
Storage Temperature:Ambient
EINECS:207-894-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together