Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
2,6-Dimethylpyrazine ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
 
2,6-Dimethylpyrazine ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    35.0 to 39.0 deg-C

 

Frequently Bought Together