Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,6-Dihydroxynaphthalene ≥95.0% (by GC)
 :  TCI
 
undefined
 
2,6-Dihydroxynaphthalene ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 95.0 %

 

 

Frequently Bought Together