
2,5-Dimethylpyrazine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:123-32-0
:
Formula:C₆H₈N₂
MDL Number:MFCD00006147
:108.14 g/mol
Boiling Pt:155 °C (1013 hPa)
Melting Pt:15 °C
Density:0,99 g/cm³ (20 °C)
Flash Pt:64°C
EINECS:204-618-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together