Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,3-Dihydroxynaphthalene ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
2,3-Dihydroxynaphthalene ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    162.0 to 165.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together