Order Entry
Netherlands
ContactUsLinkComponent
 
 
m-Tolyl isothiocyanate ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
m-Tolyl isothiocyanate ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Butylamine Method):
    min. 98.0 %

 

Frequently Bought Together