Octadecyl isocyanate ≥80.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-96-9
:
Formula:C₁₉H₃₇NO
MDL Number:MFCD00002041
Boiling Pt:220°C
Melting Pt:15…16 °C
Flash Pt:202°C
Storage Temperature:Refrigerator
EINECS:204-019-7
UN:2206
ADR:6.1,III
:295.51 g/mol
:GHS06
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 80.0 %
Specification Test Results
Frequently Bought Together