


m-Tolyl isocyanate ≥99.0% (by GC)
: TCI
:621-29-4
:
:C₈H₇NO
:MFCD00002021
:133.15 g/mol
:96°C
:195…198 °C
:1,033 g/cm³ (20 °C)
:65°C
:Refrigerator
:210-676-0
:2206
:6.1,III
:GHS05
:GHS06
:GHS08
- Appearance:Colorless to Yellow to Orange clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together