Order Entry
Netherlands
ContactUsLinkComponent
α,α,α-Trifluoro-m-tolyl isocyanate ≥96.0% (by GC)
Flammable
Toxic
Health
α,α,α-Trifluoro-m-tolyl isocyanate ≥96.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Toxic
Health
α,α,α-Trifluoro-m-tolyl isocyanate ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

Frequently Bought Together