

o-Tolyl isocyanate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:614-68-6
:
Formula:C₈H₇NO
MDL Number:MFCD00002009
Boiling Pt:186°C
Density:1,07 g/cm³ (20 °C)
Flash Pt:92°C
EINECS:210-389-0
UN:2206
ADR:6.1,III
:133.15 g/mol
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together