
3-Nitro-p-cresol ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2042-14-0
:
Formula:C₇H₇NO₃
MDL Number:MFCD00007244
:153.14 g/mol
Melting Pt:78…81 °C
EINECS:218-044-6
UN:2446
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:76.0 to 80.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together