
6-Nitro-m-cresol ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:700-38-9
:
Formula:C₇H₇NO₃
MDL Number:MFCD00007111
:153.14 g/mol
Melting Pt:56°C
Storage Temperature:Ambient
EINECS:211-843-0
UN:2446
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (Neutralization titration):min. 95.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together