


4-Nitro-m-cresol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2581-34-2
:
Formula:C₇H₇NO₃
MDL Number:MFCD00007333
Melting Pt:126 °C
Storage Temperature:Ambient
EINECS:219-952-5
UN:2446
ADR:6.1,III
:153.14 g/mol
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:127.0 to 130.0 deg-C
- Solubility in Methanol:within almost transparency
Specification Test Results
Frequently Bought Together