Order Entry
Netherlands
ContactUsLinkComponent
3-Hexyn-1-ol ≥97.0% (by GC)
Irritant
3-Hexyn-1-ol ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
3-Hexyn-1-ol ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Frequently Bought Together