
α,α,α-Trifluoro-m-cresol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:98-17-9
:
Formula:C₇H₅F₃O
MDL Number:MFCD00002299
:162.11 g/mol
Boiling Pt:178°C
Melting Pt:–2 °C
Density:1,34 g/cm³ (20 °C)
Flash Pt:73°C
Storage Temperature:Ambient
EINECS:202-645-5
ADR:8,III
:GHS05
Specification Test Results
- Purity (GC):min. 98.0 %
- Appearance:Colorless to Light orange to Yellow clear liquid
Specification Test Results
Frequently Bought Together