4'-Methylacetophenone ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:122-00-9
:
Formula:C₉H₁₀O
MDL Number:MFCD00008751
:134.18 g/mol
Boiling Pt:226°C
Melting Pt:28 °C
Density:1,013 g/cm³ (20 °C)
Flash Pt:92°C
EINECS:204-514-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together