
4'-Methoxyacetophenone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:100-06-1
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00008745
:150.18 g/mol
Boiling Pt:260°C
Melting Pt:36…38 °C
Density:1,14 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:202-815-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:36.0 to 39.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together