
2,3,5-Trifluorobenzonitrile ≥97.0% (by GC)
TCIAT2369-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
:
:157.09 g/mol
:3276
:6.1,III
:GHS06
:241154-09-6
:C₇H₂F₃N
:MFCD00083539