
4-Chlorophenyl isothiocyanate ≥98.0% (by GC)
Catalog # TCIAI0167-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:43.0 to 47.0 deg-C
Specification Test Results
About this item
:
:169.63 g/mol
Boiling Pt:127°C
Melting Pt:46 °C
Storage Temperature:Ambient
EINECS:218-358-3
UN:2811
ADR:6.1,III
:GHS06
CAS Number:2131-55-7
Formula:C₇H₄ClNS
MDL Number:MFCD00004810