



4-Chloorfenylisocyanaat ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:104-12-1
:
Formula:C₇H₄ClNO
MDL Number:MFCD00002024
:153.57 g/mol
Boiling Pt:117°C
Melting Pt:31…32 °C
Density:1,2 g/cm³ (20 °C)
Flash Pt:110 °C
EINECS:203-176-9
UN:2206
ADR:6.1,II
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Koelkast
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together