



4-Chloro-m-cresol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:59-50-7
:
Formula:C₇H₇ClO
MDL Number:MFCD00002323
Merck Index:14,02133
Boiling Pt:235°C
Melting Pt:63…65 °C
Density:0,9 g/cm³ (20 °C)
Flash Pt:118 °C
UN:3437
ADR:6.1,II
:142.58 g/mol
EINECS:200-431-6
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:64.0 to 67.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together