
5-Chloro-o-cresol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5306-98-9
:
Formula:C₇H₇ClO
MDL Number:MFCD00060672
:142.58 g/mol
Boiling Pt:225 °C (1013 hPa)
Melting Pt:75…76 °C
EINECS:226-160-3
UN:3437
ADR:6.1,II
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:72.0 to 76.0 deg-C
Specification Test Results
Frequently Bought Together