
2-Chloro-p-cresol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6640-27-3
:
Formula:C₇H₇ClO
MDL Number:MFCD00070639
Boiling Pt:196°C
Melting Pt:76 °C
Density:1,211 g/cm³ (25 °C)
Flash Pt:86°C
EINECS:229-656-8
ADR:6.1,II
:142.58 g/mol
UN:2669
:GHS06
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together