
3-Chloro-o-xylene ≥96.0% (by GC)
Catalog # TCIAC1558-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
About this item
:
:140.61 g/mol
Boiling Pt:190°C
Flash Pt:73°C
:GHS07
CAS Number:608-23-1
Formula:C₈H₉Cl
MDL Number:MFCD00060689