


4-Bromophenyl isothiocyanate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1985-12-2
:
Formula:C₇H₄BrNS
MDL Number:MFCD00004808
:214.09 g/mol
Boiling Pt:142 - 144 °C (14 -rr)
Melting Pt:60 °C
UN:1759
ADR:8,III
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:58.0 to 62.0 deg-C
Specification Test Results
Frequently Bought Together