Order Entry
Netherlands
ContactUsLinkComponent
3-Bromophenyl isothiocyanate ≥98.0% (by GC)
Corrosive
Irritant
Health
3-Bromophenyl isothiocyanate ≥98.0% (by GC)
Catalog # TCIAI0524-5G
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
Health
3-Bromophenyl isothiocyanate ≥98.0% (by GC)
Catalog # TCIAI0524-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :214.09 g/mol
Boiling Pt:256 °C (1013 hPa)
UN:1759
ADR:8,III
 :GHS05
 :GHS07
 :GHS08
CAS Number:2131-59-1
Formula:C₇H₄BrNS
MDL Number:MFCD00004803