
2-Bromo-4,6-difluorophenol ≥98.0% (by GC, titration analysis)
: TCI
:98130-56-4
:
:C₆H₃BrF₂O
:MFCD00070752
:208.99 g/mol
:53°C
:106°C
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together