

α,α,α-Trifluoro-p-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:455-14-1
:
Formula:C₇H₆F₃N
MDL Number:MFCD00064396
:161.13 g/mol
Boiling Pt:117°C
Melting Pt:3…8 °C
Density:1,3 g/cm³ (20 °C)
Flash Pt:86°C
Storage Temperature:Refrigerator
EINECS:207-236-5
UN:2810
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together