

2-Chloro-p-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:615-65-6
:
Formula:C₇H₈ClN
MDL Number:MFCD00007666
:141.6 g/mol
Boiling Pt:224°C
Melting Pt:7°C
Density:1,151 g/cm³ (20 °C)
Flash Pt:99°C
Storage Temperature:Ambient
EINECS:210-440-7
UN:3429
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together