Order Entry
Netherlands
ContactUsLinkComponent
ω,2,4-Trichloroacetophenone ≥98.0% (by GC)
Corrosive
Irritant
ω,2,4-Trichloroacetophenone ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Corrosive
Irritant
ω,2,4-Trichloroacetophenone ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    53.0 to 57.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together