
3-Fluorobenzylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:100-82-3
:
Formula:C₇H₈FN
MDL Number:MFCD00008113
:125.15 g/mol
Boiling Pt:82°C
Density:1,09 g/cm³ (20 °C)
Flash Pt:71°C
EINECS:202-891-3
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together