Order Entry
Netherlands
ContactUsLinkComponent
3,4-Dichlorobenzylamine ≥98.0% (by GC, titration analysis)
Corrosive
3,4-Dichlorobenzylamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
3,4-Dichlorobenzylamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together