
2,5-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:85118-06-5
:
Formula:C₇H₇F₂N
MDL Number:MFCD00010143
:143.14 g/mol
Boiling Pt:76 - 78 °C (15 -rr)
Density:1,22 g/cm³ (20 °C)
Flash Pt:76°C
EINECS:285-658-9
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together